Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Alkyne Agarose

Catalog No. 9019822ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-198-22ML 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-198-22ML Supplier Sigma Aldrich Supplier No. 9019822ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alkyne agarose is a 6% cross-linked agarose resin that is activated with terminal alkyne functional groups for covalent capturing of azide-tagged biomolecules. This product is adequate to work in batch or column purifications (Low Pressure).Functionalized Alkynes

Specifications

CAS 1195995-72-2
Melting Point 131°C to 133°C
Molecular Formula C31H39O3P
Quantity 250 mg
Synonym Biaryl phosphine ligand for Pd catalysis; Dicyclohexyl(2,6-dimethoxy-3-(2-methoxynaphthalen-1-yl)phenyl)phosphane
Molecular Weight (g/mol) 490.61 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.