Learn More
Medchemexpress LLC Alkyne Cholesterol | 1631985-09-5 | C26H40O | 1 MG
SDP
Catalog No. 5000372072
Encompass_Preferred
Alkyne Cholesterol is a modified lipid that functions as a click chemistry reagent containing an alkyne group. This terminal alkyne group enables highly specific linking reactions with azide-containing reagents in the presence of a copper (Cu)-containing catalyst. It can be utilized for tracking cellular cholesterol metabolism and localization.
  • Functions as a click chemistry reagent
  • Contains a terminal alkyne group
  • Facilitates specific linking reactions with azide-containing reagents
  • Requires a copper-containing catalyst
  • Used for tracking cellular cholesterol metabolism
  • Used for tracking cellular cholesterol localization
Catalog No. 50-003-72072 Supplier Medchemexpress LLC Supplier No. HY1517051MG
Alkyne Cholesterol is a modified lipid that functions as a click chemistry reagent containing an alkyne group. This terminal alkyne group enables highly specific linking reactions with azide-containing reagents in the presence of a copper (Cu)-containing catalyst. It can be utilized for tracking cellular cholesterol metabolism and localization.
  • Functions as a click chemistry reagent
  • Contains a terminal alkyne group
  • Facilitates specific linking reactions with azide-containing reagents
  • Requires a copper-containing catalyst
  • Used for tracking cellular cholesterol metabolism
  • Used for tracking cellular cholesterol localization