Learn More
Medchemexpress LLC Alkyne-cRGD | 2665674-77-9 | 99.48% | 713.78 | 5 MG
SDP
Catalog No. 5000396154
Encompass_Preferred
Alkyne-cRGD is an integrin ligand that can be used in the study of degraders. It undergoes copper-catalyzed azide-alkyne cycloaddition reaction. It is characterized by a purity of 99.48% and is supplied as a white to off-white solid.
  • Functions as an integrin ligand
  • Useful in the study of degraders
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction
  • High purity of 99.48%
  • Supplied as a white to off-white solid
Catalog No. 50-003-96154 Supplier Medchemexpress LLC Supplier No. HYP64385MG
Alkyne-cRGD is an integrin ligand that can be used in the study of degraders. It undergoes copper-catalyzed azide-alkyne cycloaddition reaction. It is characterized by a purity of 99.48% and is supplied as a white to off-white solid.
  • Functions as an integrin ligand
  • Useful in the study of degraders
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction
  • High purity of 99.48%
  • Supplied as a white to off-white solid