Learn More
LICORbio IRDye 800CW Alkyne Infrared Dye (0.5 mg)
Catalog No. NC1001434
Encompass_Preferred
IRDye 800CW Alkyne provides the functionality for preferential labeling of molecules containing azide groups (–N3) with IRDye 800CW.
Catalog No. NC1001434 Supplier LICORbio Supplier No. 92960002
IRDye 800CW Alkyne provides the functionality for preferential labeling of molecules containing azide groups (–N3) with IRDye 800CW.