Learn More
Glen Research Alkyne-Modifier Serinol Phosphoramidite, 10-1992-02, C42H55N4O7P, M.W. 758.88, 5' terminus or internal modification, Click Chemistry application, 3 min coupling time, -10 to -30 °C storage
Catalog No. NC3342834
Encompass_Preferred
Alkyne-Modifier Serinol Phosphoramidite, 10-1992-02, C42H55N4O7P, M.W. 758.88, 5' terminus or internal modification, Click Chemistry application, 3 min coupling time, -10 to -30 °C storage
Catalog No. NC3342834 Supplier Glen Research Supplier No. 10199202
Alkyne-Modifier Serinol Phosphoramidite, 10-1992-02, C42H55N4O7P, M.W. 758.88, 5' terminus or internal modification, Click Chemistry application, 3 min coupling time, -10 to -30 °C storage