Learn More
Medchemexpress LLC Alkyne-P60 | 99.9% | 2087.47 | 1 MG
SDP
Catalog No. 5000391912
Encompass_Preferred
Alkyne-P60 is a potent 15-mer peptide inhibitor of Foxp3. It binds with Foxp3, hinders its nuclear translocation, and diminishes Foxp3-mediated inhibition of NFKB and NFAT functions. Alkyne-P60 is a ligand for target protein for PROTAC.
  • Potent 15-mer peptide inhibitor of Foxp3.
  • Binds with Foxp3, hinders nuclear translocation, and diminishes Foxp3-mediated inhibition of NFKB and NFAT functions.
  • Ligand for target protein for PROTAC.
  • For research use only.
Catalog No. 50-003-91912 Supplier Medchemexpress LLC Supplier No. HY1629461MG
Alkyne-P60 is a potent 15-mer peptide inhibitor of Foxp3. It binds with Foxp3, hinders its nuclear translocation, and diminishes Foxp3-mediated inhibition of NFKB and NFAT functions. Alkyne-P60 is a ligand for target protein for PROTAC.
  • Potent 15-mer peptide inhibitor of Foxp3.
  • Binds with Foxp3, hinders nuclear translocation, and diminishes Foxp3-mediated inhibition of NFKB and NFAT functions.
  • Ligand for target protein for PROTAC.
  • For research use only.