Learn More
Medchemexpress LLC Alkyne-PEG2-iodide | 1234387-33-7 | C7H11IO2 | 1 G
SDP
Catalog No. 5000373439
Encompass_Preferred
Alkyne-PEG2-iodide is a click chemistry reagent with terminal alkyne and alkyl iodide groups.
  • Click chemistry reagent
  • Features terminal alkyne groups
  • Features alkyl iodide groups
  • Targets ADC linker
  • Pathway is antibody-drug conjugate/ADC related
Catalog No. 50-003-73439 Supplier Medchemexpress LLC Supplier No. HY1516651G
Alkyne-PEG2-iodide is a click chemistry reagent with terminal alkyne and alkyl iodide groups.
  • Click chemistry reagent
  • Features terminal alkyne groups
  • Features alkyl iodide groups
  • Targets ADC linker
  • Pathway is antibody-drug conjugate/ADC related