Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Alkyne-Peg5-Acid

Catalog No. 76416725MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-416-725MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 76-416-725MG Supplier Sigma Aldrich Supplier No. 76416725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alkyne-PEG5-acid may be used in the synthesis of peptide nucleic acid (PNA) oligomers that can conjugate with vitamin B12 via Cu-catalyzed 1,3-dipolar cycloaddition in order to study if vitamin B12 to transport PNA oligonucleotides into bacterial cells.

Specifications

CAS 157141-27-0
Melting Point 119°C to 125°C
Molecular Formula C10H17NO5
Linear Formula C10H17NO5
Quantity 1 g
MDL Number MFCD23135644
Molecular Weight (g/mol) 231.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.