Learn More
Medchemexpress LLC Alkyne-phenol (alkyne tyramide) | 1694495-59-4 | 99.7% | 217.26 g/mol | C13H15NO2 | 5 MG
SDP
Catalog No. 5000262936
Encompass_Preferred
Alkyne-phenol is a clickable alkyne-containing probe (alkyne tyramide) used for APEX2-mediated proximity labeling and bioorthogonal tagging. It facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAC) for downstream conjugation and is supplied at high purity for reliable research applications.
  • Contains an alkyne functional group enabling CuAAC click chemistry.
  • Optimized for APEX2 proximity labeling in intact cells.
  • Improves cell wall permeability compared with biotin-phenol.
  • High chemical purity supports reproducible results.
  • Available in solid sizes and solution for experimental flexibility.
Catalog No. 50-002-62936 Supplier Medchemexpress LLC Supplier No. HY1314425MG
Alkyne-phenol is a clickable alkyne-containing probe (alkyne tyramide) used for APEX2-mediated proximity labeling and bioorthogonal tagging. It facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAC) for downstream conjugation and is supplied at high purity for reliable research applications.
  • Contains an alkyne functional group enabling CuAAC click chemistry.
  • Optimized for APEX2 proximity labeling in intact cells.
  • Improves cell wall permeability compared with biotin-phenol.
  • High chemical purity supports reproducible results.
  • Available in solid sizes and solution for experimental flexibility.