Learn More
Medchemexpress LLC Alkyne-SNAP | 1104822-07-2 | C18H18N6O2 | 1 MG
SDP
Catalog No. 5000373214
Encompass_Preferred
Alkyne-SNAP is an alkyne-conjugated benzylguanine. Its benzylguanine moiety reacts with the SNAP-tag, enabling irreversible and covalent labeling of SNAP fusion proteins with an additional alkyne functionality for subsequent click chemistry conjugation.
  • Alkyne-conjugated benzylguanine
  • Enables irreversible and covalent labeling of SNAP fusion proteins
  • Provides alkyne functionality for click chemistry conjugation
  • White to off-white solid appearance
  • For research use only
  • Purity of 99.32%
Catalog No. 50-003-73214 Supplier Medchemexpress LLC Supplier No. HY1516451MG
Alkyne-SNAP is an alkyne-conjugated benzylguanine. Its benzylguanine moiety reacts with the SNAP-tag, enabling irreversible and covalent labeling of SNAP fusion proteins with an additional alkyne functionality for subsequent click chemistry conjugation.
  • Alkyne-conjugated benzylguanine
  • Enables irreversible and covalent labeling of SNAP fusion proteins
  • Provides alkyne functionality for click chemistry conjugation
  • White to off-white solid appearance
  • For research use only
  • Purity of 99.32%