Learn More
Medchemexpress LLC HY-131442 50mg Medchemexpress, Alkyne tyramide CAS:1694495-59-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13144250MG
Encompass_Preferred
Medchemexpress, HY-131442 50mg Alkyne tyramide CAS:1694495-59-4 Alkyne tyramide is a clickable ascorbate peroxidase 2 (APEX2) probe. Alkyne tyramide substantially improves APEX-labeling efficiency in intact yeast cells, as it is more cell wall-permeant than APEX2 substrate biotin-phenol (BP). Alkyne tyramide also facilitates the identification of APEX-labeling sites, allowing the unambiguous assignment of membrane topology of mitochondrial proteins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0258