Learn More
Selleck Chemical LLC Alkyne tyramide-E0471-5MG
SDP
Catalog No. 503137623
Encompass_Preferred
Alkyne tyramide is a clickable ascorbate peroxidase 2 (APEX2) probe substantially improves APEX-labeling efficiency in intact yeast cells also facilitates the identification of APEX-labeling sites allowing the unambiguous assignment of membrane topology of mitochondrial proteins
Catalog No. 50-313-7623 Supplier Selleck Chemical LLC Supplier No. E04715MG
Alkyne tyramide is a clickable ascorbate peroxidase 2 (APEX2) probe substantially improves APEX-labeling efficiency in intact yeast cells also facilitates the identification of APEX-labeling sites allowing the unambiguous assignment of membrane topology of mitochondrial proteins