Learn More
Cayman Chemical Alkynyl-biotin, Purity: ≥95%, CAS: 1011268-28-2, 5 mg
SDP
Catalog No. 502955931
Encompass_Preferred
A form of biotin with a terminal alkyne group separated by two polyethylene glycol moieties; used to add biotin to other molecules bearing an azide group using click chemistry
Catalog No. 50-295-5931 Supplier Cayman Chemical Supplier No. 130385 MG
A form of biotin with a terminal alkyne group separated by two polyethylene glycol moieties; used to add biotin to other molecules bearing an azide group using click chemistry