Learn More
TARGETMOL CHEMICALS INC Alkynyl Palmitic Acid 1MG
SDP
Catalog No. 502879625
Encompass_Preferred
Also available in 1 mL, 5 mg, 50 mg and bulk. Please contact Fisher for quotes. Alkynyl Palmitic Acid is an alkyl chain-based PROTAC linker that can be used in PROTAC synthesis.
Catalog No. 50-287-9625 Supplier TARGETMOL CHEMICALS INC Supplier No. T141851MG
Also available in 1 mL, 5 mg, 50 mg and bulk. Please contact Fisher for quotes. Alkynyl Palmitic Acid is an alkyl chain-based PROTAC linker that can be used in PROTAC synthesis.