Learn More
Medchemexpress LLC HY-W040304 1mg Medchemexpress, Alkynyl Palmitic Acid CAS:99208-90-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0403041MG
Encompass_Preferred
Medchemexpress, HY-W040304 1mg Alkynyl Palmitic Acid CAS:99208-90-9 Alkynyl Palmitic Acid is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3471