Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alkynyl palmitic acid | 99208-90-9 | 97.0% | 252.39 g/mol | C16H28O2 | 25 MG
SDP

Catalog No. 5000208865
Encompass_Preferred

Alkynyl palmitic acid is an alkyne-functionalized palmitic acid used as a click-chemistry reagent and as an alkyl-chain PROTAC linker. It contains a terminal alkyne that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) to conjugate to azide-containing molecules. The material is supplied as a solid with defined storage recommendations and high purity.

  • Alkyne-functionalized palmitic acid for CuAAC click reactions
  • Used as an alkyl-chain PROTAC linker in chemical synthesis
  • Purity 97.0%
  • Solid form, white to off-white
  • Storage: powder -20°C for 3 years; in solvent -80°C for 6 months, -20°C for 1 month
  • CAS number 99208-90-9; molecular weight 252.39 g/mol
  • SMILES C#CCCCCCCCCCCCCCC(O)=O

Catalog No. 50-002-08865 Supplier Medchemexpress LLC Supplier No. 25MGHYW040304
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Alkynyl palmitic acid is an alkyne-functionalized palmitic acid used as a click-chemistry reagent and as an alkyl-chain PROTAC linker. It contains a terminal alkyne that enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) to conjugate to azide-containing molecules. The material is supplied as a solid with defined storage recommendations and high purity.

  • Alkyne-functionalized palmitic acid for CuAAC click reactions
  • Used as an alkyl-chain PROTAC linker in chemical synthesis
  • Purity 97.0%
  • Solid form, white to off-white
  • Storage: powder -20°C for 3 years; in solvent -80°C for 6 months, -20°C for 1 month
  • CAS number 99208-90-9; molecular weight 252.39 g/mol
  • SMILES C#CCCCCCCCCCCCCCC(O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.