Learn More
Anton Paar All the chemistry you need
Catalog No. 5000580508
Encompass_Preferred
Cora 100 has inbuilt chemical intelligence. Spectral databases stored in the Raman analyzer quickly identify hazardous materials, narcotics, explosives, and chemical warfare agents. The result gives you more than a cryptic chemical name, it also tells you whether a substance is listed or regulated, or a known precursor of such a compound. Additionally, CAS numbers and safety data are provided. For increased customization you can easily create your own personal library and add new spectra if required.
Catalog No. 50-005-80508 Supplier Anton Paar Supplier No. 220967
Please contact your sales representative for a quote or further ordering instructions.
Cora 100 has inbuilt chemical intelligence. Spectral databases stored in the Raman analyzer quickly identify hazardous materials, narcotics, explosives, and chemical warfare agents. The result gives you more than a cryptic chemical name, it also tells you whether a substance is listed or regulated, or a known precursor of such a compound. Additionally, CAS numbers and safety data are provided. For increased customization you can easily create your own personal library and add new spectra if required.