Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

all-cis-4,7,10,13,16-Docosapentaenoic acid, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013716 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3716 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3716 Supplier MilliporeSigma Supelco Supplier No. 1856610MG
Add to Cart
Add to Cart

Docosapentaenoic acid is a very long-chain polyunsaturated fatty acid, and has two or more double bonds in its acyl chains.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 25182-74-5
Physical Form Neat
Linear Formula C22H34O2
MDL Number MFCD00216034
Synonym (4 Z,7 Z,10 Z,13 Z,16 Z)-4,7,10,13,16-Docosapentaenoic acid; (all-Z)-4,7,10,13,16-Docosapentaenoic acid; Osbond acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 330.50
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.