Learn More
Shawmut Protex™ All-Day-Comfort™ N95 Particulate Respirators
GSA_VASDP
Catalog No. 17000342
Click to view available options
Model:
SR9520-12
SR9520S-12
Size:
Medium/Large
Small
Strap Attachment Type:
Two-Strap Design
Welded Two Strap Design
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size Strap Attachment Type
17-000-342 SR9520S-12 Small Welded Two Strap Design
17-448-400 SR9520-12 Medium/Large Two-Strap Design
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-000-342 Supplier Shawmut Protex™ Supplier No. SR9520S12
Add to Cart
Add to Cart
This item is not returnable. View return policy
Made in the USA, molded-cup N95 respirator is easier to breathe through while offering a comfortable and secure fit for maximum protection and less eyeglass fogging. Meets ASTM F3407 Respirator Fit Capability standard to securely fit a wide range of face sizes and shapes.
Made by Shawmut Corp™, this NIOSH-approved, disposable N95 particulate respirator meets NIOSH 42 CFR 84 N95 requirements for a minimum 95% filtration efficiency. Its patent-pending All-Day Comfort™ System includes a soft, non-abrasive inner shell precisely thermoformed for a superior fit; a lightweight, super-high-efficiency filter layer for easier breathing, a comfortable viscoelastic nose foam for a secure seal, and expanded force-curve elastic head straps that are engineered to fit a wider range of sizes.
  • Molded cup style N95 for superior seal and protection
  • 6
  • Better breathability and cooler, fresher air inside the cup with Proprietary Shawmut MB™ filtering technology
  • Meets ASTM F3407 Respirator Fit Capability Standard to fit widest range of face sizes and shapes
  • Provides respiratory protection by removing more than 95% of airborne particles larger than 0.3 microns
  • Secure fit and tight seal around user's face ensures the respirator will perform at N95 standards while being comfortable to wear for long periods
  • Less eyeglass fogging with unique viscoelastic nose foam and aluminum nose clip
  • Two-strap welded design for more secure fit
  • Ideal for sanding, cutting, sawing, yard work, painting, grinding or other heavy industries
  • Also appropriate for general masking and travel recommendations, as well as liquid or non-oil-based particles from sprays that do not also emit oil aerosols or vapors
  • Made in the USA
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size Small
With Exhalation Valve No
Filter Efficiency 95%
Filtration Layer Material Meltblown Polypropylene
Latex Free Yes
Material Headstrap Elastic
Model SR9520S-12
Nose Piece Material Aluminum
Nosepiece Type Adjustable Nose Clip
Quantity 20 Pk.
Sterility Non-sterile
Strap Attachment Type Welded Two Strap Design
FDA Cleared Surgical Mask No
Show More Show Less
Not made with natural rubber latex.