Learn More
Wells Lamont™ All Day™ Sleeve
GSA_VAGREENER_CHOICE
Catalog No. 17000482 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
14 in.
18 in.
24 in.
Style:
Sleeve with Thumb Hole
Standard Sleeve
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
17-000-482 14 in. Standard Sleeve
17-000-483 18 in. Standard Sleeve
17-000-485 - Sleeve with Thumb Hole
17-000-484 - Standard Sleeve
17-000-486 24 in. Sleeve with Thumb Hole
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-000-482 Supplier Wells Lamont™ Supplier No. AD14
Add to Cart
Add to Cart
The All Day™ Sleeve offers ANSI A2 cut resistance and is made out of recycled water bottles. Features an inner cotton liner for comfort, elastic on both ends, and is latex free. Made in the USA.
  • Protect arms and the environment with cut resistant sleeves made of recycled material that would normally go to the landfill
  • High strength All Day™ fiber provides superior snag, tear, and abrasion resistance
  • Free of harsh dyes, chemicals, and latex
  • Lightweight construction provides all day comfort and fit
  • Elastic on both ends hold sleeve in place and cotton inner layer for added comfort
  • Three lengths are available depending on the length needed for protection
  • Available with or without a thumb hole
  • Not intended for use around sparks and flames
  • Made in the USA
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Cut-resistant Sleeve
Size 14 in.
Material Recycled Water Bottles
Sterility Non-sterile
Style Standard Sleeve
Disposable Single-use
Cut Resistance Rating ANSI 2, ANSI A2
Latex Free Yes
No. per Case 144/Cs.