Learn More
Electron Microscopy Sciences All Glass Nebulizer Only
Supplier: Electron Microscopy Sciences 7050605
An all glass unit for the simple production of microdroplets. An object is held vertically in front of the nebulizer outlet and by squeezing the atomizer a fine spray is created. The nebulizer set comes with an All Glass Nebulizer bulb and Atomizer.