Learn More
Electron Microscopy Sciences All Glass Nebulizer Set
SDP
Supplier:  Electron Microscopy Sciences 7050501
Encompass
An all glass unit for the simple production of microdroplets. An object is held vertically in front of the nebulizer outlet and by squeezing the atomizer a fine spray is created. The nebulizer set comes with an All Glass Nebulizer bulb and Atomizer.
Catalog No. 50-930-365