Learn More
Electron Microscopy Sciences All Glass Nebulizer Set
SDP
Catalog No. 50930366
Encompass
An all glass unit for the simple production of microdroplets. An object is held vertically in front of the nebulizer outlet and by squeezing the atomizer a fine spray is created. The nebulizer set comes with an All Glass Nebulizer bulb and Atomizer.
Catalog No. 50-930-366 Supplier Electron Microscopy Sciences Supplier No. 7050505
Add to Cart
Add to Cart
Request a Quote
An all glass unit for the simple production of microdroplets. An object is held vertically in front of the nebulizer outlet and by squeezing the atomizer a fine spray is created. The nebulizer set comes with an All Glass Nebulizer bulb and Atomizer.