Learn More
Mac-Mod All-Guard Cartridge Holder (AKA H0001 & HI161): requires HI-081 PEEK coupler
SDP
Catalog No. NC1239226
Encompass
This guard cartridge holder promotes increased column lifetime. Improved hardware. Higher pressure rating. Low internal swept volume. This guard cartridge holder is supplied in new hardware, leading to more robust and higher efficiency separations. Chromatographic selectivity will be unaffected by this change.
Catalog No. NC1239226 Supplier Mac-Mod Supplier No. 80101N
Add to Cart
Add to Cart
This guard cartridge holder promotes increased column lifetime. Improved hardware. Higher pressure rating. Low internal swept volume. This guard cartridge holder is supplied in new hardware, leading to more robust and higher efficiency separations. Chromatographic selectivity will be unaffected by this change.