Learn More
Apexbio Technology LLC HyperScribe™ All in One mRNA Synthesis Kit Plus 2 (ARCA, 5-moUTP, T7, poly(A)), 25rxns
SDP
Catalog No. 501998234
Encompass_Preferred
HyperScribe™ All in One mRNA Synthesis Kit Plus 2 (ARCA, 5-moUTP, T7, poly(A)) is designed for quick production of ARCA capped, 5-moUTP modified, poly(A) tailed mRNA in vitro. Capped mRNA is synthesized by co-transcriptional incorporation of Anti-Reverse Cap Analog (ARCA, Catalog No. B8175) using T7 RNA Polymerase. The capping of ARCA ensures high translation efficacy. The addition of 5-moUTP (Catalog No. B8061) can suppress RNA-mediated innate immune activation. After a brief DNase I treatment to remove the template DNA, capped and modified mRNA is poly(A) tailed with Poly(A) Polymerase. Polyadenylation (i.e. addition of Poly(A) tail) plays an important role in the stabilization of RNA in eukaryotes and enhances the efficiency of translation initiation.
Catalog No. 50-199-8234 Supplier Apexbio Technology LLC Supplier No. K106525
HyperScribe™ All in One mRNA Synthesis Kit Plus 2 (ARCA, 5-moUTP, T7, poly(A)) is designed for quick production of ARCA capped, 5-moUTP modified, poly(A) tailed mRNA in vitro. Capped mRNA is synthesized by co-transcriptional incorporation of Anti-Reverse Cap Analog (ARCA, Catalog No. B8175) using T7 RNA Polymerase. The capping of ARCA ensures high translation efficacy. The addition of 5-moUTP (Catalog No. B8061) can suppress RNA-mediated innate immune activation. After a brief DNase I treatment to remove the template DNA, capped and modified mRNA is poly(A) tailed with Poly(A) Polymerase. Polyadenylation (i.e. addition of Poly(A) tail) plays an important role in the stabilization of RNA in eukaryotes and enhances the efficiency of translation initiation.