Learn More
Mopec All-In-One Tookit
SDP
Catalog No. 23900750 Shop All Mopec Products
Change view
Click to view available options
Width (English):
48 in.
60 in.
72 in.
96 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
23-900-750 48 in.
23-900-751 60 in.
23-900-752 72 in.
23-900-753 96 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-900-750 Supplier Mopec Supplier No. BU001F48
Add to Cart
Add to Cart
These toolkits have been created with the expertise and experience of Pathologists’ Assistants to address the basic needs in every gross room.
The Toolkit includes:
  • (4) Scalpel Blades, Sterile #22, 100 pack
  • (4) Scalpel Blades, Sterile #60, 100 pack
  • (4) BladeFlask EVO Universal Blade Remover
  • (3) Premium Biopsy Bags, Small, 1000 pack
  • (2) BxPads, Blue Sponge, 100 pack
  • (2) BxWraps, 2 x 3 in., 500 pack
  • (2) Formaway Formalin. Neutralizing Pads, 8 x 11 in., 100 pack
  • (2) Formaway Formalin. Neutralizing Pads, 11 x 15 in., 100 pack
  • (1) SaniPath Disinfectant Foaming Spray
  • (1) EazySoak Instrument Cleaner
  • (1) ClearSteel Stainless Steel Cleaner and Polish Spray
  • (1) PathOdorizer Plus, Deodorizer, Cleaner and Degreaser
Specifications
Product Type Dissection Tool Kit
Width (English) 48 in.