Learn More
BURKLE INC All-layer drill, V4A, length 59.06 in. (150 cm), volume 8.79 oz. (260 ml)
Catalog No. 5000025588
Encompass_Preferred
The all-layer drill offers an excellent solution to the challenge of taking samples from highly compacted materials. Its specially designed drill bits make it possible to penetrate the medium and sample the entire depth without much effort or time. This is especially valuable for high-density media and compacted powders and granules that are difficult to sample using conventional methods.
The all-layer auger is constructed of high-quality materials that ensure long life and efficiency when sampling from large containers, mixing tanks and big bags in a variety of industries.
The all-layer auger provides representative sampling through all layers, even with highly compressed media.
Catalog No. 50-000-25588 Supplier BURKLE INC Supplier No. 53100150
The all-layer drill offers an excellent solution to the challenge of taking samples from highly compacted materials. Its specially designed drill bits make it possible to penetrate the medium and sample the entire depth without much effort or time. This is especially valuable for high-density media and compacted powders and granules that are difficult to sample using conventional methods.
The all-layer auger is constructed of high-quality materials that ensure long life and efficiency when sampling from large containers, mixing tanks and big bags in a variety of industries.
The all-layer auger provides representative sampling through all layers, even with highly compressed media.