Learn More
BURKLE INC All-layer scratcher, V4A, Ø 0.98 in (25 mm), 39.37 in (100 cm), volume 6.1 oz (180 ml)
Catalog No. 5000026098
Encompass_Preferred
The sample lance scraper is particularly suitable for sampling non-free-flowing, coagulating powders and granulates. The scraper edge efficiently guides the material into a sealable inner tube for precise, contamination-free sampling. Variable zones within the inner tube can be individually adjusted by sliding PTFE separators to prevent mixing of samples and allow precise separation of different layers or areas. The sample lance is ideal for removing large volumes of sample from different layers without mixing materials.
Two additional handles allow for safe insertion, fixation, and rotation of the material, and easy disassembly of the instrument facilitates thorough cleaning.
Catalog No. 50-000-26098 Supplier BURKLE INC Supplier No. 53511000
The sample lance scraper is particularly suitable for sampling non-free-flowing, coagulating powders and granulates. The scraper edge efficiently guides the material into a sealable inner tube for precise, contamination-free sampling. Variable zones within the inner tube can be individually adjusted by sliding PTFE separators to prevent mixing of samples and allow precise separation of different layers or areas. The sample lance is ideal for removing large volumes of sample from different layers without mixing materials.
Two additional handles allow for safe insertion, fixation, and rotation of the material, and easy disassembly of the instrument facilitates thorough cleaning.