Learn More
Cole-Parmer Oakton Buffer Pack; 60 mL of each pH 1.68, 4.01, 7.00, 10.01, and 12.46
Catalog No. 501961282 Shop All Cole-Parmer Products
Encompass_Preferred
• Oakton pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards • If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days • Store buffers away from direct sunlight.
Catalog No. 50-196-1282 Supplier Cole-Parmer Supplier No. 0065314
• Oakton pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations • Manufactured under ISO 9000 quality guidelines and traceable to NIST standards • If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days • Store buffers away from direct sunlight.