Learn More
Cole-Parmer Thermo Scientific Nalgene 4150-2000 All-plastic Dewar flask, 2 L
Catalog No. NC9713625 Shop All Cole-Parmer Products
Encompass_Preferred
Implosion proof and safe to use with LN2, dry ice, solvent mixtures, and hot fluids. Double-wall construction, flasks are made of high-density polyethylene (HDPE); annulus is filled with insulating CFC-free urethane foam. Metal tabs keep lid securely in place. Broad-base design reduces the chance of accidental spills. Include a vented, insulated HDPE cover, molded in ribs, securing tabs, and pour spout.
This item is not returnable. View return policy
Catalog No. NC9713625 Supplier Cole-Parmer Supplier No. 0672640
Implosion proof and safe to use with LN2, dry ice, solvent mixtures, and hot fluids. Double-wall construction, flasks are made of high-density polyethylene (HDPE); annulus is filled with insulating CFC-free urethane foam. Metal tabs keep lid securely in place. Broad-base design reduces the chance of accidental spills. Include a vented, insulated HDPE cover, molded in ribs, securing tabs, and pour spout.