Learn More
Nexcelom Bioscience Llc Cellometer Chambers 900/CS
Catalog No. NC0057157
Encompass
Case of 900 slides good for 1,800 counts. Disposable, all-plastic cell counting chambers for Cellometer cell counters. 20 uL sample size. Disposable, no cross contamination or cleaning. Standard thickness chamber accommodating cells from 2 to 80um in diameter. Each slide has protective film on both sides that must be removed before use. Case contains 12 boxes of 75 slides. Expiration date is 1 year from date of manufacture. Check expiration date on outside of cardboard box. Compatible with all Cellometer instruments.
Catalog No. NC0057157 Supplier Nexcelom Bioscience Llc Supplier No. CHT4SD100014/DUP
Case of 900 slides good for 1,800 counts. Disposable, all-plastic cell counting chambers for Cellometer cell counters. 20 uL sample size. Disposable, no cross contamination or cleaning. Standard thickness chamber accommodating cells from 2 to 80um in diameter. Each slide has protective film on both sides that must be removed before use. Case contains 12 boxes of 75 slides. Expiration date is 1 year from date of manufacture. Check expiration date on outside of cardboard box. Compatible with all Cellometer instruments.