Learn More
Brady™ All Purpose Cable Lockout Without Cable
Catalog No. 19103936 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
19-103-936 Red
1 options
Catalog No. 19-103-936 Supplier Brady™ Supplier No. 50944
  • $63.50
    $45.65 / Each of 1
    Save $17.85
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Be prepared for any lockout situation. The Brady™ All Purpose Cable Lockout is highly versatile, making it perfect for all of your unusual energy isolating devices that are difficult to lock out.
  • Incredibly versatile
  • Perfect for all of your unusal energy isolating devices that are difficult to lock out
  • Compact, lightweight body is made of a rugged impact modified glass filled nylon
  • Superior chemical, corrosion and temperature resistant properties
  • 8 ft., 12 ft., 16 ft. and 20 ft. cables are sold separately
Specifications
Product Type Pull-tight Cable Lockout Device
Color Red
Material Glass-filled Nylon
Dimensions (L x W x H) 3 x 6.5 x 1.1 in.
Quantity 1 Ea.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.