Learn More
MSC Detco All-Purpose Cleaner, Liquid, Pine Scent, 55 gal
Catalog No. 502819423
Encompass
This all-purpose cleaner is a liquid, water-based, dilutable concentrate supplied in a 55-gallon drum. It features a pine scent, is non-acid, biodegradable, and designed for use on washable surfaces.
  • Liquid form
  • Drum container type
  • Capacity: 55 gal
  • Pine scent
  • Non-disinfectant
  • Suitable for washable surfaces
  • Water-based solution
  • Dilutable concentrate
  • Non-acid formula
  • Biodegradable
Catalog No. 50-281-9423 Supplier MSC Supplier No. 33703000
This all-purpose cleaner is a liquid, water-based, dilutable concentrate supplied in a 55-gallon drum. It features a pine scent, is non-acid, biodegradable, and designed for use on washable surfaces.
  • Liquid form
  • Drum container type
  • Capacity: 55 gal
  • Pine scent
  • Non-disinfectant
  • Suitable for washable surfaces
  • Water-based solution
  • Dilutable concentrate
  • Non-acid formula
  • Biodegradable