Learn More
MSC Super Clean Cleaner-Degreaser, Industrial Strength, All-Purpose, 55 gal
Catalog No. 502867369
Encompass
Super Clean Cleaner-Degreaser is an industrial-strength cleaning solution formulated to effectively dissolve tough grease, grime, oil, wax, dirt, and tar. It offers versatile cleaning for various surfaces, capable of tackling both heavy-duty and everyday messes.
  • Dissolves grease and removes grime, oil, wax, dirt, and tar
  • Can be used at full strength for powerful cleaning or diluted for lighter tasks
  • Biodegradable and phosphate-free formulation
  • Effectively fights stubborn grease and oil residue
  • Removes scuff marks and stains without damaging surfaces
Catalog No. 50-286-7369 Supplier MSC Supplier No. 14084057
Super Clean Cleaner-Degreaser is an industrial-strength cleaning solution formulated to effectively dissolve tough grease, grime, oil, wax, dirt, and tar. It offers versatile cleaning for various surfaces, capable of tackling both heavy-duty and everyday messes.
  • Dissolves grease and removes grime, oil, wax, dirt, and tar
  • Can be used at full strength for powerful cleaning or diluted for lighter tasks
  • Biodegradable and phosphate-free formulation
  • Effectively fights stubborn grease and oil residue
  • Removes scuff marks and stains without damaging surfaces