Learn More
MSC Betco Disinfectant Cleaner, RTU, Citrus Floral, 32 oz, 12/Carton
Catalog No. 502866966
Encompass
This ready-to-use disinfectant cleaner provides broad-spectrum efficacy against bacteria and viruses on hard, non-porous surfaces. It is suitable for use in various institutional and commercial settings including healthcare, educational, and food service facilities.
  • Kills over 99.9% of bacteria and viruses within 3 to 5 minutes contact time
  • Effective against SARS-CoV-2 on hard non-porous surfaces
  • Formulated with botanical active ingredient and citric acid, free from harsh chemicals
  • Can be used as a food contact sanitizer with no rinsing or wiping required
  • Non-irritating, with no harsh fumes
  • Complies with OSHA's bloodborne pathogen standard
  • Will not scratch or bleach surfaces
Catalog No. 50-286-6966 Supplier MSC Supplier No. 35154202
This ready-to-use disinfectant cleaner provides broad-spectrum efficacy against bacteria and viruses on hard, non-porous surfaces. It is suitable for use in various institutional and commercial settings including healthcare, educational, and food service facilities.
  • Kills over 99.9% of bacteria and viruses within 3 to 5 minutes contact time
  • Effective against SARS-CoV-2 on hard non-porous surfaces
  • Formulated with botanical active ingredient and citric acid, free from harsh chemicals
  • Can be used as a food contact sanitizer with no rinsing or wiping required
  • Non-irritating, with no harsh fumes
  • Complies with OSHA's bloodborne pathogen standard
  • Will not scratch or bleach surfaces