Learn More
MSC CRC HydroForce All Purpose Degreaser, Spray Bottle, 946 mL
Catalog No. 502866527
Encompass
HydroForce All Purpose Degreaser is an aqueous-based all-purpose degreaser that effectively breaks up and lifts off grease, oil, and dirt deposits. It is versatile enough for all degreasing tasks, ideal for facilities seeking an improvement over current aqueous degreasers.
  • Moderate pH (9.5) and non-corrosive
  • Biodegradable
  • Non-abrasive
  • Non-flammable aqueous formulation
  • Low VOC content
  • NSF C1 registered
  • No Class I or II ozone depleting chemicals
  • Safe on most plastics
  • Ready-to-use
Catalog No. 50-286-6527 Supplier MSC Supplier No. 02451045
HydroForce All Purpose Degreaser is an aqueous-based all-purpose degreaser that effectively breaks up and lifts off grease, oil, and dirt deposits. It is versatile enough for all degreasing tasks, ideal for facilities seeking an improvement over current aqueous degreasers.
  • Moderate pH (9.5) and non-corrosive
  • Biodegradable
  • Non-abrasive
  • Non-flammable aqueous formulation
  • Low VOC content
  • NSF C1 registered
  • No Class I or II ozone depleting chemicals
  • Safe on most plastics
  • Ready-to-use