Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences All Purpose Reagent Reservoirs, 48mL
SDP

Catalog No. 03410916 Shop All DWK Life Sciences Products
Change view
Click to view available options
Channels:
8
12
No. per Pack:
30
100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Channels No. per Pack
03-410-916 8 30
03-410-917 8 100
03-410-918 12 30
03-410-919 12 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-410-916 Supplier DWK Life Sciences Supplier No. W845900
Add to Cart
Add to Cart

Well suited for single and multichannel pipetting.

  • Made from polystyrene (PE)
  • Available in 8-channel and 12-channel formats
  • Packs of 30 or 100
TRUSTED_SUSTAINABILITY

Specifications

Material Polystyrene
Type Reagent Reservoir
No. per Pack 30
Volume (Metric) Well 48 mL
Channels 8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.