Learn More
Description
- Made from polystyrene (PS)
- Available in 8-channel and 12-channel formats
- Non-sterile configuration
- 48mL capacity
- Available in packs of 30 or 100
Specifications
Specifications
| Capacity (Metric) | 48 mL |
| Material | Polystyrene |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| No. per Pack | 100 |
| Volume (Metric) Well | 48 mL |
| Autoclavable | Not Autoclavable |
| Channels | 12 |
| Disposable | Single-use |
| Bottom Shape | Convoluted |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
