Learn More
Learning Resources All Ready for First Grade Readiness Kit

Description
Ocean-themed at-home educational kit to prepare children for first grade
Targets key first-grade skills including word building, writing, addition and subtraction, telling time, etc.
Includes:
- 36-page journal
- 20 double-sided story starter cards
- 20 ocean counters
- 10 double-sided word problem cards
- 6 word family cubes
- 6 measuring fish
- 1 write-and-wipe clock
- 24-page write-and-wipe activity book
- parent guide
- storage box
Specifications
Specifications
| Grade Level | Elementary |
| Grade | 1 |
| Class Size | 1 Class |
| Product Type | First Grade Readiness Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.