Learn More
Learning Resources All Ready For Preschool Readiness Kit

Description
Comprehensive kit to prepare children for preschool
Targets essential preschool skills including counting, alphabet, color identification, and shape recognition
Features a family theme
Includes:
- assortment of colorful manipulatives
- write-and-wipe activity book for children
- 32-page guide for parents
- storage box
Specifications
Specifications
| Grade Level | Early Childhood |
| Grade | Pre-K |
| Class Size | 1 Class |
| Product Type | Preschool Readiness Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.