Learn More
Learning Resources All Ready for Kindergarten Readiness Kit

Description
Comprehensive kit to prepare children for kindergarten
Teach counting, addition and subtraction, word building, handwriting, and beginning reading
Features the 'In the Garden' classroom theme and includes an expansive guide for parents.
Specifications
Specifications
| Grade Level | Elementary |
| Grade | K |
| Class Size | 1 Class |
| Product Type | Kindergarten Readiness Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.