Learn More
Felix Storch Inc All-Refrigerator/All-Freezer Combination 28" Wide Performance
SDP
Catalog No. NC3901238
Encompass_Preferred
The ARFG18PVDLPW features an upper refrigerator compartment with a small window and independent manual defrost freezer below. Both sections include adjustable plastic-coated wire shelves that can be positioned at 1/2 inch increments to accommodate virtually any sized item. This unit includes LED internal lighting with an on/off rocker switch. The interior is constructed from white powder coated metal to ensure added durability and better temperature retention. The ARFG18PVDLPW features a dual microprocessor temperature controller (refrigerator range: +2 to +8ºC; freezer range: -30 to -10ºC). The buffered temperature probes are encased in glycol-filled bottles to better simulate temperature of the stored product, with an additional sensor that reads air temperature to ensure superior temperature control. This unit ships with PharmaWatch DLPWRF, a wireless environmental temperature monitoring device (ETM) that enables cloud-based data management.
Catalog No. NC3901238 Supplier Felix Storch Inc Supplier No. ARFG18PVDLPWLHD
Add to Cart
Add to Cart
Request a Quote
The ARFG18PVDLPW features an upper refrigerator compartment with a small window and independent manual defrost freezer below. Both sections include adjustable plastic-coated wire shelves that can be positioned at 1/2 inch increments to accommodate virtually any sized item. This unit includes LED internal lighting with an on/off rocker switch. The interior is constructed from white powder coated metal to ensure added durability and better temperature retention. The ARFG18PVDLPW features a dual microprocessor temperature controller (refrigerator range: +2 to +8ºC; freezer range: -30 to -10ºC). The buffered temperature probes are encased in glycol-filled bottles to better simulate temperature of the stored product, with an additional sensor that reads air temperature to ensure superior temperature control. This unit ships with PharmaWatch DLPWRF, a wireless environmental temperature monitoring device (ETM) that enables cloud-based data management.