Learn More
TARGETMOL CHEMICALS INC ALL-TRANS-4-OXORETINOIC 5MG
SDP
Catalog No. 502579194
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. all-trans-4-Oxoretinoic acid (all-trans 4-Keto Retinoic Acid) is an active metabolite of vitamin A. purity: 96%
Catalog No. 50-257-9194 Supplier TARGETMOL CHEMICALS INC Supplier No. T74085MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. all-trans-4-Oxoretinoic acid (all-trans 4-Keto Retinoic Acid) is an active metabolite of vitamin A. purity: 96%