Learn More
Medchemexpress LLC All-trans-4-oxoretinoic acid | 38030-57-8 | MFCD00870472 | 98.0% | 314.42 g·mol⁻¹ | C20H26O3 | 1 ML
SDP
Catalog No. 5000241262
Encompass_Preferred
All-trans-4-oxoretinoic acid is an active metabolite of vitamin A that binds nuclear retinoic acid receptors (RARs) to induce gene transcription. It is provided as a 10 mM solution in DMSO for use in biochemical and cell-based research; handle and store under recommended cold conditions.
  • Active metabolite that modulates RAR-mediated gene transcription.
  • Supplied as a 10 mM solution in DMSO for convenient dosing.
  • High purity for research applications (≈98.0%).
  • Chemical formula C20H26O3 and molecular weight 314.42 g·mol⁻¹.
  • Store in solvent at -80°C for optimal long-term stability.
Catalog No. 50-002-41262 Supplier Medchemexpress LLC Supplier No. HY107494A10MM 1ML
All-trans-4-oxoretinoic acid is an active metabolite of vitamin A that binds nuclear retinoic acid receptors (RARs) to induce gene transcription. It is provided as a 10 mM solution in DMSO for use in biochemical and cell-based research; handle and store under recommended cold conditions.
  • Active metabolite that modulates RAR-mediated gene transcription.
  • Supplied as a 10 mM solution in DMSO for convenient dosing.
  • High purity for research applications (≈98.0%).
  • Chemical formula C20H26O3 and molecular weight 314.42 g·mol⁻¹.
  • Store in solvent at -80°C for optimal long-term stability.