Learn More
Medchemexpress LLC HY-W004500 100mg Medchemexpress, All-trans-retinal CAS:116-31-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW004500100MG
Encompass_Preferred
Medchemexpress, HY-W004500 100mg All-trans-retinal CAS:116-31-4 All-trans-retinal is a one of the major vitamin A metabolites in the retina. In physiological conditions, all-trans-RAL is regenerated to the visual chromophore, 11-cis-retinal. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4483