Learn More
Apexbio Technology LLC all-trans Retinal,250mg CAS# 116-31-4
SDP
Supplier:  Apexbio Technology LLC C4061250
Encompass_Preferred
All-trans Retinal, also known as Vitamin A aldehyde or Retinaldehyde, is one of the many forms of vitamin A and also the oxidation product of all-trans retinol [1]. All-trans Retinal are associated with one of the two isoforms of cellular retinol-binding proteins (CRBP-I and CRBP-II) with Kd values of 50 and 90 nM, respectively [1].CRBP-I and CRBP-II were the first intracellular retinoid-binding proteins. Both proteins display a similar binding affinity towards retinal. They play important roles in retinoid biology and regulation of the metabolism of retinol and retinal. CRBP-I is used to regulate vitamin A storage and synthesis of retinoic acid. And CRBP-II has a role in the initial processing of retinol from food [1].Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0328