Learn More
BIOGEMS INTERNATIONAL INC All-Trans Retinoic Acid 1g
SDP
Catalog No. 501530001
Encompass
Retinoic Acid is vitamin A derivative that is an agonist for the retinoic acid receptors, which act as transcription factors that regulate the growth and differentiation of normal and malignant cells. It is reported to promote the differentiation of embryonic cells into motor neurons, pancreatic progenitors and cardiomyocytes.
Catalog No. 50-153-0001 Supplier BIOGEMS INTERNATIONAL INC Supplier No. 30279491G
Retinoic Acid is vitamin A derivative that is an agonist for the retinoic acid receptors, which act as transcription factors that regulate the growth and differentiation of normal and malignant cells. It is reported to promote the differentiation of embryonic cells into motor neurons, pancreatic progenitors and cardiomyocytes.