Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC all-trans-Retinol | 68-26-8 | 98.3% | 25 MG
SDP

Catalog No. 5000413827
Encompass_Preferred

Vitamin A (Vitamin A1) is an endogenous metabolite for research use only. It is not sold to patients. This compound is unstable in solutions, therefore freshly prepared solutions are recommended.

  • Solid appearance, off-white to yellow color
  • Molecular weight: 286.45
  • Formula: C20H30O
  • Store at -20°C, protect from light, stored under nitrogen
  • Soluble in ethanol (100 mg/mL)

Catalog No. 50-004-13827 Supplier Medchemexpress LLC Supplier No. 25MGHYB1342
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vitamin A (Vitamin A1) is an endogenous metabolite for research use only. It is not sold to patients. This compound is unstable in solutions, therefore freshly prepared solutions are recommended.

  • Solid appearance, off-white to yellow color
  • Molecular weight: 286.45
  • Formula: C20H30O
  • Store at -20°C, protect from light, stored under nitrogen
  • Soluble in ethanol (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.