Learn More
Hach Company All Weather Refrig Sampler
Catalog No. 5000207543 Shop All Hach Company Products
Encompass
AS950 All Weather Refrigerated Sampler-230 V-Heater-2 Sensor Port Options-Discrete (24x) 1 L Bottle Kit-Digital pH SensorIncludes1 ea AS950 controller with sensor ports on 230V All Weather base wtih controller compartment heater1 ea Bottle kit (AW241000) includes 24- 1L poly bottles w/caps (737)-bottle tray (1511)-retainer (1322)-and distributor arm (8841)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)1 ea digital pH sensor (DPD2P1)1 ea adapter cable (9501200)
Catalog No. 50-002-07543 Supplier Hach Company Supplier No. ASA.CSXX4X413X
Add to Cart
Add to Cart
Request a Quote
AS950 All Weather Refrigerated Sampler-230 V-Heater-2 Sensor Port Options-Discrete (24x) 1 L Bottle Kit-Digital pH SensorIncludes1 ea AS950 controller with sensor ports on 230V All Weather base wtih controller compartment heater1 ea Bottle kit (AW241000) includes 24- 1L poly bottles w/caps (737)-bottle tray (1511)-retainer (1322)-and distributor arm (8841)1 ea 25 vinyl intake tubing (920)1 ea strainer (926)1 ea digital pH sensor (DPD2P1)1 ea adapter cable (9501200)