Learn More
Hach Company AS950 All-Weather Refrigerated Sampler, 115 V, 20 L PE Bottle
Catalog No. 5000207779 Shop All Hach Company Products
Encompass
The AS950 All-Weather Refrigerated Sampler provides reliable refrigerated composite and discrete sample collection for harsh outdoor environments. It maintains sample temperature with an air-sensing thermostat and supports intuitive programming and data transfer for simplified field operation.
  • Refrigerated sample chamber with air-sensing thermostat that maintains temperature per USEPA and international guidelines.
  • Large full-color display with single-screen programming for intuitive operation.
  • USB upload/download and program copying between samplers for convenient data transfer.
  • Durable, corrosion-resistant base and controller housing for long-term outdoor use.
  • Easy maintenance with spring-mounted rollers and replaceable pump tubing and desiccant.
  • 20 L polyethylene sample bottle capacity and NEMA 6 / IP68 controller housing for field durability.
Catalog No. 50-002-07779 Supplier Hach Company Supplier No. ASA.CXXX1X11XX
Add to Cart
Add to Cart
Request a Quote
The AS950 All-Weather Refrigerated Sampler provides reliable refrigerated composite and discrete sample collection for harsh outdoor environments. It maintains sample temperature with an air-sensing thermostat and supports intuitive programming and data transfer for simplified field operation.
  • Refrigerated sample chamber with air-sensing thermostat that maintains temperature per USEPA and international guidelines.
  • Large full-color display with single-screen programming for intuitive operation.
  • USB upload/download and program copying between samplers for convenient data transfer.
  • Durable, corrosion-resistant base and controller housing for long-term outdoor use.
  • Easy maintenance with spring-mounted rollers and replaceable pump tubing and desiccant.
  • 20 L polyethylene sample bottle capacity and NEMA 6 / IP68 controller housing for field durability.